(±)-4-Methyl-1,3-dioxolane ≥98.0%
Supplier: TCI America
Synonyms:
4-Methyl-1,3-dioxolane
M0916-25ML
TCM0916-025MLEA
98.42
USD
TCM0916-025ML
(±)-4-Methyl-1,3-dioxolane ≥98.0%
(±)-4-Methyl-1,3-dioxolane
CAS Number: 1072-47-5
MDL Number: MFCD00014109
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 85
Flash Point (°C): -2
Specific Gravity (20/20): 0.99
Formula:
C₄H₈O₂ MW: 88.11 g/mol Boiling Pt: 85 °C Density: 0.99 (20 °C) Flash Pt: –2 °C |
MDL Number:
MFCD00014109 CAS Number: 1072-47-5 EINECS: 214-008-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...