(R)-(+)-2-(ɑ-Methylbenzylamino)-5-nitropyridine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (R)-5-Nitro-N-(1-phenylethyl)pyridin-2-amine

M0909-5G
TCM0909-005GEA 224 USD
TCM0909-005G
(R)-(+)-2-(ɑ-Methylbenzylamino)-5-nitropyridine ≥98.0% (by titrimetric analysis)
(R)-(+)-2-(alpha-Methylbenzylamino)-5-nitropyridine

CAS Number: 64138-65-4
MDL Number: MFCD00060067
Molecular Formula: C13H13N3O2
Molecular Weight: 243.27
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 83
Specific rotation [a]20/D: 146.5 deg (C=2, CHCl3)

Formula: C₁₃H₁₃N₃O₂
MW: 243.27 g/mol
Melting Pt: 83 °C
MDL Number: MFCD00060067
CAS Number: 64138-65-4
https://us.vwr.com/stibo/web/std.lang.all/44/45/28974445.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR