(±)-3-Methyl-2-pentanol ≥95.0%
Supplier: TCI America
Synonyms:
3-Methyl-2-pentanol
M0850-5ML
M0850-25ML
TCM0850-025MLEA
260.55
USD
TCM0850-025ML
TCM0850-005ML
(±)-3-Methyl-2-pentanol ≥95.0%
(±)-3-Methyl-2-pentanol
CAS Number: 565-60-6
MDL Number: MFCD00004528
Molecular Formula: C6H14O
Molecular Weight: 102.18
Purity/Analysis Method:
Form: Clear Liquid
Flash Point (°C): 40
Specific Gravity (20/20): 0.83
Formula:
C₆H₁₄O MW: 102.18 g/mol Boiling Pt: 134 °C (1013 hPa) Melting Pt: –85 °C Density: 0.83 (20 °C) Flash Pt: 40 °C |
MDL Number:
MFCD00004528 CAS Number: 565-60-6 EINECS: 209-281-6 UN: 1987 ADR: 3,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...