2-Methyl-2-butenenitrile ≥70.0%
Supplier: TCI America
Synonyms:
2-Cyano-2-butene
, 1-Methyl-1-propenyl Cyanide
M0839-500ML
M0839-25ML
TCM0839-025MLEA
38.55
USD
TCM0839-025ML
TCM0839-500ML
2-Methyl-2-butenenitrile ≥70.0%
2-Methyl-2-butenenitrile
CAS Number: 4403-61-6
MDL Number: MFCD00037169
Molecular Formula: C5H7N
Molecular Weight: 81.12
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 50
Flash Point (°C): -17
Specific Gravity (20/20): 0.82
Other Product Names / Synonyms: 2-Cyano-2-butene, 1-Methyl-1-propenyl Cyanide
Formula:
C₅H₇N MW: 81.12 g/mol Boiling Pt: 50 °C Density: 0.82 (20 °C) Flash Pt: –17 °C |
MDL Number:
MFCD00037169 CAS Number: 4403-61-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...