2-Methoxyphenothiazine ≥98.0% (by HPLC)

Supplier: TCI America

M0822-25G
TCM0822-025GEA 203 USD
TCM0822-025G
2-Methoxyphenothiazine ≥98.0% (by HPLC)
2-Methoxyphenothiazine

CAS Number: 1771-18-2
MDL Number: MFCD00037838
Molecular Formula: C13H11NOS
Molecular Weight: 229.30
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 188

Formula: C₁₃H₁₁NOS
MW: 229.30 g/mol
Melting Pt: 188 °C
MDL Number: MFCD00037838
CAS Number: 1771-18-2
https://us.vwr.com/stibo/web/std.lang.all/42/92/28974292.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR