(4S,5S)-(-)-4-Methoxymethyl-2-methyl-5-phenyl-2-oxazoline ≥95.0% (by GC, titration analysis)

M0803-1G
TCM0803-001GEA 150.96 USD
TCM0803-001G
(4S,5S)-(-)-4-Methoxymethyl-2-methyl-5-phenyl-2-oxazoline ≥95.0% (by GC, titration analysis)
(4S,5S)-(-)-4-Methoxymethyl-2-methyl-5-phenyl-2-oxazoline

CAS Number: 52075-14-6
MDL Number: MFCD00078100
Molecular Formula: C12H15NO2
Molecular Weight: 205.26
Purity/Analysis Method: 95.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.07
Specific rotation [a]20/D: -108 deg (C=10, CHCl3)

Formula: C₁₂H₁₅NO₂
Density: 1,07 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00078100
CAS Number: 52075-14-6

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR