2-(Methylsulfonyl)acetophenone ≥98.0%

Supplier: TCI America

Synonyms: 2-(Methylsulfonyl)-1-phenylethanone2-(Methylsulphonyl)-1-phenylethanone

M0712-5G
TCM0712-005GEA 173.52 USD
TCM0712-005G
2-(Methylsulfonyl)acetophenone ≥98.0%
2-(Methylsulphonyl)acetophenone

CAS Number: 3708-04-1
MDL Number: MFCD00025078
Molecular Formula: C9H10O3S
Molecular Weight: 198.24
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 108


Other Product Names / Synonyms: Benzoylmethyl Methyl Sulfone, Methyl Phenacyl Sulfone

Formula: C₉H₁₀O₃S
Melting Pt: 108 °C
MDL Number: MFCD00025078
CAS Number: 3708-04-1
https://us.vwr.com/stibo/web/std.lang.all/41/88/28974188.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR