2-Methyl-4-ethylthiazoline ≥88.0%

Supplier: TCI America

Synonyms: 4-Ethyl-2-methylthiazoline

M0689-25ML
TCM0689-025MLEA 241.49 USD
TCM0689-025ML
2-Methyl-4-ethylthiazoline ≥88.0%
2-Methyl-4-ethylthiazoline

CAS Number: 4293-61-2
MDL Number: MFCD00015468
Molecular Formula: C6H11NS
Molecular Weight: 129.22
Purity/Analysis Method: 88.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: 4-Ethyl-2-methylthiazoline

Formula: C₆H₁₁NS
MW: 129.22 g/mol
Density: 1.00 (20 °C)
MDL Number: MFCD00015468
CAS Number: 4293-61-2
https://us.vwr.com/stibo/web/std.lang.all/41/65/28974165.jpg

Order Now

Specification Test Results

Purity/Analysis Method 88.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR