Methylfluorone Hemisulfate

Supplier: TCI America

Synonyms: 9-Methyl-2,3,7-trihydroxy-6-fluorone Hemisulphate9-Methyl-2,3,7-trihydroxy-6-fluorone Hemisulfate

M0678-1G
TCM0678-001GEA 100.21 USD
TCM0678-001G
Methylfluorone Hemisulfate
Methylfluorone Hemisulfate

[for Detection of Sb,Ge,Mo,Sn]
MDL Number: MFCD00022274
Molecular Formula: C14H10O5
Molecular Weight: 307.27
Form: Crystal
Color: Yellow


Other Product Names / Synonyms: 9-Methyl-2,3,7-trihydroxy-6-fluorone Hemisulfate

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR