N-Methyl-bis-trifluoroacetamide ≥97.0%
Supplier: TCI America
|
Danger
|
Synonyms:
MBTFA
M0671-5ML
M0671-1ML
TCM0671-001MLEA
61.9
USD
TCM0671-001ML
TCM0671-5ML
N-Methyl-bis-trifluoroacetamide ≥97.0%
N-Methylbistrifluoroacetamide
[Trifluoroacylating Agent]
CAS Number: 685-27-8
MDL Number: MFCD00000412
Molecular Formula: C5H3F6NO2
Molecular Weight: 223.07
Purity/Analysis Method:
Form: Clear Liquid
Flash Point (°C): 45
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Other Fluorinated Group Introducing Agents, Analytical Science, Acylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: N,N-Bis(trifluoroacetyl)methylamine, MBTFA
Formula:
C₅H₃F₆NO₂ MW: 223.07 g/mol Flash Pt: 45 °C |
MDL Number:
MFCD00000412 CAS Number: 685-27-8 EINECS: 211-680-5 UN: 2920 ADR: 8/3,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...