Isopentylbenzene ≥98.0%

Supplier: TCI America

Synonyms: Isoamylbenzene(3-Methylbutyl)benzene

M0656-5ML M0656-25ML
TCM0656-005MLEA 90.3 USD
TCM0656-005ML TCM0656-025ML
Isopentylbenzene ≥98.0%
Isopentylbenzene

CAS Number: 2049-94-7
MDL Number: MFCD00008948
Molecular Formula: C11H16
Molecular Weight: 148.25
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 199
Flash Point (°C): 65
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: Isoamylbenzene

Formula: (C₆H₅)CH₂CH₂CH(CH₃)₂
MW: 148.25 g/mol
Boiling Pt: 199 °C
Density: 0.86 (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00008948
CAS Number: 2049-94-7
EINECS: 218-075-5
https://us.vwr.com/stibo/web/std.lang.all/41/17/28974117.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR