5-Methyl-2-hexanone oxime ≥97.0%

Synonyms: Isoamyl Methyl Ketoxime

M0643-25ML
TCM0643-025MLEA 109.14 USD
TCM0643-025ML
5-Methyl-2-hexanone oxime ≥97.0%
5-Methyl-2-hexanone oxime

CAS Number: 624-44-2
MDL Number: MFCD00013934
Molecular Formula: C7H15NO
Molecular Weight: 129.20
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: Isoamyl Methyl Ketoxime

Formula: C₇H₁₅NO
MW: 129.20 g/mol
Density: 0.89 (20 °C)
MDL Number: MFCD00013934
CAS Number: 624-44-2
https://us.vwr.com/stibo/web/std.lang.all/41/08/28974108.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR