Monooctyl maleate ≥95.0%

Synonyms: Maleic Acid Mono(2-ethylhexyl) EsterMono(2-ethylhexyl) MaleateMaleic Acid Monooctyl Ester

M0615-25ML M0615-500ML
TCM0615-500MLEA 158.1 USD
TCM0615-500ML TCM0615-025ML
Monooctyl maleate ≥95.0%
Monooctyl maleate

CAS Number: 2370-71-0
MDL Number: MFCD00151510
Molecular Formula: C12H20O4
Molecular Weight: 228.29
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 137
Specific Gravity (20/20): 1.04
Storage Temperature: 0°C


Product Categories: Materials Chemistry, Copolymerization Monomers
Other Product Names / Synonyms: Maleic Acid Mono(2-ethylhexyl) Ester, Mono(2-ethylhexyl) Maleate, Maleic Acid Monooctyl Ester

Formula: C₁₂H₂₀O₄
MW: 228.29 g/mol
Density: 1.04 (20 °C)
Flash Pt: 137 °C
MDL Number: MFCD00151510
CAS Number: 2370-71-0
https://us.vwr.com/stibo/web/std.lang.all/40/76/28974076.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR