Ethyl-4-[(4-methoxybenzylidene)amino]cinnamate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-(p-Anisalamino)cinnamic Acid Ethyl Ester4-[(p-Anisylidene)amino]cinnamic Acid Ethyl EsterEthyl 4-(p-Anisalamino)cinnamateEthyl 4-[(p-Anisylidene)amino]cinnamate4-[(4-Methoxybenzylidene)amino]cinnamic Acid Ethyl Ester

M0602-1G
TCM0602-001GEA 499 USD
TCM0602-001G
Ethyl-4-[(4-methoxybenzylidene)amino]cinnamate ≥98.0% (by titrimetric analysis)
Ethyl-4-[(4-methoxybenzylidene)amino]cinnamate

CAS Number: 6421-30-3
MDL Number: MFCD00059231
Molecular Formula: C19H19NO3
Molecular Weight: 309.37
Purity/Analysis Method: 98.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)

Formula: C₁₉H₁₉NO₃
MW: 309.37 g/mol
MDL Number: MFCD00059231
CAS Number: 6421-30-3
https://us.vwr.com/stibo/web/std.lang.all/40/61/28974061.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR