Melissic acid ≥95.0%

Supplier: TCI America

Synonyms: Triacontanoic acid

M0595-100MG M0595-1G
TCM0595-001GEA 409.05 USD
TCM0595-001G TCM0595-100MG
Melissic acid ≥95.0%
Mellissic acid

CAS Number: 506-50-3
MDL Number: MFCD00002813
Molecular Formula: C30H60O2
Molecular Weight: 452.81
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Melting point (°C): 93


Product Categories: Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Triacontanoic Acid

Formula: C₃₀H₆₀O₂
MW: 452.81 g/mol
Melting Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00002813
CAS Number: 506-50-3
EINECS: 208-042-3
https://us.vwr.com/stibo/web/std.lang.all/40/54/28974054.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR