2-Methylbutyl-DL-2-methylbutyrate ≥97.0%

Supplier: TCI America

Synonyms: DL-2-Methylbutyric Acid 2-Methylbutyl Ester

M0578-25ML
TCM0578-025MLEA 25.36 USD
TCM0578-025ML
2-Methylbutyl-DL-2-methylbutyrate ≥97.0%
2-Methylbutyl-DL-2-methylbutyrate

CAS Number: 2445-78-5
MDL Number: MFCD00059395
Molecular Formula: C10H20O2
Molecular Weight: 172.27
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 187
Flash Point (°C): 67
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: DL-2-Methylbutyric Acid 2-Methylbutyl Ester

Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Boiling Pt: 187 °C
Density: 0.86 (20 °C)
Flash Pt: 67 °C
MDL Number: MFCD00059395
CAS Number: 2445-78-5
https://us.vwr.com/stibo/web/std.lang.all/40/36/28974036.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR