Ethyl acetylsalicylate ≥98.0%
Supplier: TCI America
Synonyms:
Acetylsalicylic Acid Ethyl Ester
, 2-Acetoxybenzoic Acid Ethyl Ester
, Ethyl 2-Acetoxybenzoate
CAS Number: 529-68-0
MDL Number: MFCD00026824
Molecular Formula: C11H12O4
Molecular Weight: 208.21
Purity/Analysis Method:
Form: Clear Liquid
Flash Point (°C): 107
Specific Gravity (20/20): 1.15
Other Product Names / Synonyms: 2-Acetoxybenzoic Acid Ethyl Ester, Ethyl 2-Acetoxybenzoate, Acetylsalicylic Acid Ethyl Ester
Formula:
C₁₁H₁₂O₄ MW: 208.21 g/mol Density: 1.15 (20 °C) Flash Pt: 107 °C |
MDL Number:
MFCD00026824 CAS Number: 529-68-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...