Calcium methacrylate hydrate
Supplier: TCI America
Synonyms:
Methacrylic Acid Calcium Salt
M0548-25G
TCM0548-025GEA
124.78
USD
TCM0548-025G
Calcium methacrylate hydrate
Calcium methacrylate hydrate
CAS Number: 16809-88-4
MDL Number: MFCD00151661
Molecular Formula: C4H6O2
Molecular Weight: 210.24
Form: Crystal
Color: White
Other Product Names / Synonyms: Methacrylic Acid Calcium Salt
Formula:
C₄H₆O₂ MW: 210.24 g/mol |
MDL Number:
MFCD00151661 CAS Number: 16809-88-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...