TOKYM0527

M0527-25G M0527-5G
TCM0527-25GEA 290.18 USD
TCM0527-25G TCM0527-005G
TOKYM0527
CAS Number: 70-25-7
MDL Number: MFCD00007034
Molecular Formula: C2H5N5O3
Molecular Weight: 147.09
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Storage Temperature: 0-10°C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR