Methylene blue hydrate ≥70.0% (by gravimetric analysis)

Supplier: TCI America
Warning

Synonyms: Methylthioninium chloride 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride

M0501-25G
TCM0501-025GEA 39.63 USD
TCM0501-025G
Methylene blue hydrate ≥70.0% (by gravimetric analysis)
Methylene blue hydrate

CAS Number: 61-73-4
MDL Number: MFCD00012111
Molecular Formula: C16H18ClN3S
Molecular Weight: 319.85
Purity/Analysis Method: 70.0% (W)
Form: Crystal
Lambda max.: 663 nm (EtOH(50vol%))


Product Categories: Bioscience, Stains & Dyes
Other Product Names / Synonyms: 3,7-Bis(dimethylamino)-5-phenothiazinium Chloride, Basic Blue 9

Formula: C₁₆H₁₈ClN₃S
MW: 319.86 g/mol (anhy)
MDL Number: MFCD00150006
CAS Number: 61-73-4
EINECS: 200-515-2
https://us.vwr.com/stibo/web/std.lang.all/39/62/28973962.jpg

Order Now

Specification Test Results

Purity/Analysis Method 70.0% (W)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR