Ethyl-N-methylcarbamate ≥98.0%

Supplier: TCI America

Synonyms: N-Methylcarbamic Acid Ethyl EsterEthyl methylcarbamateN-Methylurethane

M0456-25G
TCM0456-025GEA 166.41 USD
TCM0456-025G
Ethyl-N-methylcarbamate ≥98.0%
Ethyl-N-methylcarbamate

CAS Number: 105-40-8
MDL Number: MFCD00041924
Molecular Formula: C4H9NO2
Molecular Weight: 103.12
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 170
Flash Point (°C): 61
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: N-Methylurethane, N-Methylcarbamic Acid Ethyl Ester

Formula: C₄H₉NO₂
MW: 103.12 g/mol
Boiling Pt: 170 °C
Density: 1.02 (20 °C)
Flash Pt: 61 °C
MDL Number: MFCD00041924
CAS Number: 105-40-8
https://us.vwr.com/stibo/web/std.lang.all/39/00/28973900.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR