Diethyl methyloxalacetate ≥96.0%

Supplier: TCI America

Synonyms: Ethyl 2-EthoxalylpropionateDiethyl 2-methyl-3-oxosuccinateMethyloxalacetic Acid Diethyl Ester2-Ethoxalylpropionic Acid Ethyl Ester

M0381-25G
TCM0381-025GEA 194.88 USD
TCM0381-025G
Diethyl methyloxalacetate ≥96.0%
Diethyl methyloxalacetate

CAS Number: 759-65-9
MDL Number: MFCD00009163
Molecular Formula: C9H14O5
Molecular Weight: 202.21
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 133
Flash Point (°C): 97
Specific Gravity (20/20): 1.09


Other Product Names / Synonyms: Methyloxalacetic Acid Diethyl Ester, 2-Ethoxalylpropionic Acid Ethyl Ester, Ethyl 2-Ethoxalylpropionate

Formula: C₉H₁₄O₅
MW: 202.21 g/mol
Boiling Pt: 133 °C
Density: 1.09 (20 °C)
Flash Pt: 97 °C
MDL Number: MFCD00009163
CAS Number: 759-65-9
https://us.vwr.com/stibo/web/std.lang.all/38/03/28973803.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR