Methyl octyl sulfide ≥98.0%

Supplier: TCI America

M0379-25ML
TCM0379-025MLEA 171.46 USD
TCM0379-025ML
Methyl octyl sulfide ≥98.0%
Methyl octyl sulphide

CAS Number: 3698-95-1
MDL Number: MFCD00039990
Molecular Formula: C9H20S
Molecular Weight: 160.32
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 218
Specific Gravity (20/20): 0.85

https://us.vwr.com/stibo/web/std.lang.all/38/02/28973802.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR