1-(4-Nitrophenyl)-3-methyl-5-pyrazolone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Methyl-1-(4-nitrophenyl)-5-pyrazolone5-Methyl-2-(4-nitrophenyl)-2,4-dihydro-3H-pyrazol-3-one

M0377-25G
TCM0377-025GEA 317.56 USD
TCM0377-025G
1-(4-Nitrophenyl)-3-methyl-5-pyrazolone ≥98.0% (by GC, titration analysis)
1-(4-Nitrophenyl)-3-methyl-5-pyrazolone

CAS Number: 6402-09-1
MDL Number: MFCD00020751
Molecular Formula: C10H9N3O3
Molecular Weight: 219.20
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 222


Other Product Names / Synonyms: 3-Methyl-1-(4-nitrophenyl)-5-pyrazolone

Formula: C₁₀H₉N₃O₃
MW: 219.20 g/mol
Melting Pt: 222 °C
MDL Number: MFCD00020751
CAS Number: 6402-09-1
https://us.vwr.com/stibo/web/std.lang.all/38/00/28973800.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR