Methylmagnesium Iodide 33% ∼2 mol/L in ethyl ether
M0364-100G
TCM0364-100GEA
58.83
USD
TCM0364-100G
Methylmagnesium Iodide 33% ∼2 mol/L in ethyl ether
Methylmagnesium Iodide
CAS Number: 917-64-6
MDL Number: MFCD00001026
Molecular Formula: CH3IMg
Molecular Weight: 166.24
Flash Point (°C): -45
Formula:
CH₃IMg Flash Pt: –45 °C |
MDL Number:
MFCD00001026 CAS Number: 917-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...