Ethyl-3-Amino-2-methylbenzoate ≥98.0% (by GC, titration analysis)

Synonyms: Ethyl 3-Amino-o-toluate3-Amino-2-methylbenzoic Acid Ethyl EsterEthyl 3-amino-2-methylbenzoate3-Amino-o-toluic Acid Ethyl Ester

A1574-25G A1574-5G
TCA1574-25GEA 581.95 USD
TCA1574-25G TCA1574-5G
Ethyl-3-Amino-2-methylbenzoate ≥98.0% (by GC, titration analysis)
Ethyl-3-Amino-2-methylbenzoate

CAS Number: 57414-85-4
MDL Number: MFCD00957246
Molecular Formula: C10H13NO2
Molecular Weight: 179.22
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 150
Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: 3-Amino-2-methylbenzoic Acid Ethyl Ester, Ethyl 3-Amino-o-toluate, 3-Amino-o-toluic Acid Ethyl Ester

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Boiling Pt: 150 °C
Density: 1.11 (20 °C)
MDL Number: MFCD00957246
CAS Number: 57414-85-4
https://us.vwr.com/stibo/web/std.lang.all/96/88/28929688.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR