2-Methyl-1,5-heptadiene (cis- and trans- mixture)

Supplier: TCI America

M0316-5ML
TCM0316-005MLEA 152 USD
TCM0316-005ML
2-Methyl-1,5-heptadiene (cis- and trans- mixture)
2-Methyl-1,5-heptadiene (cis- and trans- mixture)

CAS Number: 6766-54-7
MDL Number: MFCD00059240
Molecular Formula: C8H14
Molecular Weight: 110.20
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 118
Flash Point (°C): 14
Specific Gravity (20/20): 0.74

Formula: C₈H₁₄
MW: 110.20 g/mol
Boiling Pt: 118 °C
Density: 0,74 (20 °C)
Flash Pt: 14 °C
MDL Number: MFCD00059240
CAS Number: 6766-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR