3-Methyl-1,3-pentadiene ≥99.0%

M0314-5ML
TCM0314-005MLEA 335.78 USD
TCM0314-005ML
3-Methyl-1,3-pentadiene ≥99.0%
3-Methyl-1,3-pentadiene

CAS Number: 4549-74-0
MDL Number: MFCD00009277
Molecular Formula: C6H10
Molecular Weight: 82.15
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 77
Flash Point (°C): -28
Specific Gravity (20/20): 0.73
Storage Temperature: 0-10°C

Formula: C₆H₁₀
Boiling Pt: 77 °C
Density: 0.73 (20 °C)
Flash Pt: –28 °C
MDL Number: MFCD00009277
CAS Number: 4549-74-0
UN: 2461
ADR: 3,II

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR