5-Methylnonane ≥99.0%

Supplier: TCI America

M0284-5ML
TCM0284-005MLEA 734.54 USD
TCM0284-005ML
5-Methylnonane ≥99.0%
5-Methylnonane

CAS Number: 15869-85-9
MDL Number: MFCD00048832
Molecular Formula: C10H22
Molecular Weight: 142.29
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 165
Flash Point (°C): 38
Specific Gravity (20/20): 0.73

Formula: C₁₀H₂₂
Boiling Pt: 165 °C
Density: 0,73 (20 °C)
Flash Pt: 38 °C
MDL Number: MFCD00048832
CAS Number: 15869-85-9

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR