2-Mercapto-5-thiazolidone ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2-Thio-5-thiazolidone
M0263-1G
TCM0263-001GEA
208
USD
TCM0263-001G
2-Mercapto-5-thiazolidone ≥98.0% (by HPLC, titration analysis)
2-Mercapto-5-thiazolidone
CAS Number: 6913-23-1
MDL Number: MFCD00059745
Molecular Formula: C3H3NOS2
Molecular Weight: 133.18
Purity/Analysis Method:
Form: Crystal
Storage Temperature:
Other Product Names / Synonyms: 2-Thio-5-thiazolidone
Formula:
C₃H₃NOS₂ MW: 133.18 g/mol |
MDL Number:
MFCD00059745 CAS Number: 6913-23-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...