Methylenedisalicylic acid (mixture of isomers) ≥90.0% (by titrimetric analysis)
Supplier: TCI America
M0221-25G
M0221-500G
TCM0221-025GEA
17.24
USD
TCM0221-025G
TCM0221-500G
Methylenedisalicylic acid (mixture of isomers) ≥90.0% (by titrimetric analysis)
Methylenedisalicylic acid (mixture of isomers)
CAS Number: 27496-82-8
MDL Number: MFCD00016506
Molecular Formula: C15H12O6
Molecular Weight: 288.26
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 245
Formula:
C₁₅H₁₂O₆ Melting Pt: 245 °C |
MDL Number:
MFCD00016506 CAS Number: 27496-82-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...