Mercaptoacetic acid (thioglycolic acid) ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
Mercaptoacetic acid
CAS Number: 68-11-1
MDL Number: MFCD00004876
Molecular Formula: C2H4O2S
Molecular Weight: 92.11
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 96
Melting point (°C): -16
Flash Point (°C): 119
Specific Gravity (20/20): 1.33
Product Categories: Synthetic Organic Chemistry, Carboxylic Acids (Multi-component Reaction), Bioscience, Polyamine Synthesis Reagents, Crosslinkers
Other Product Names / Synonyms: Mercaptoacetic Acid
Formula:
HSCH₂COOH MW: 92.12 g/mol Boiling Pt: 96 °C Melting Pt: –16 °C Density: 1.33 (20 °C) Flash Pt: 119 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004876 CAS Number: 68-11-1 EINECS: 200-677-4 UN: 1940 ADR: 8,II Merck Index: 12,09472 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...