Dimethyl-DL-malate ≥97.0%
Supplier: TCI America
Synonyms:
DL-Apple Acid Dimethyl Ester
, DL-Malic Acid Dimethyl Ester
M0025-10G
TCM0025-010GEA
103.5
USD
TCM0025-010G
Dimethyl-DL-malate ≥97.0%
Dimethyl-DL-malate
CAS Number: 1587-15-1
MDL Number: MFCD00025868
Molecular Formula: C6H10O5
Molecular Weight: 162.14
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 205
Specific Gravity (20/20): 1.24
Other Product Names / Synonyms: DL-Apple Acid Dimethyl Ester, DL-Malic Acid Dimethyl Ester
Formula:
C₆H₁₀O₅ MW: 162.14 g/mol Boiling Pt: 205 °C Density: 1.24 (20 °C) |
MDL Number:
MFCD00025868 CAS Number: 1587-15-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...