Ethyl lignocerate ≥98.0%

Supplier: TCI America

Synonyms: Lignoceric Acid Ethyl EsterEthyl TetracosanoateTetracosanoic Acid Ethyl Ester

L0111-5G
TCL0111-005GEA 325 USD
TCL0111-005G
Ethyl lignocerate ≥98.0%
Ethyl lignocerate

CAS Number: 24634-95-5
MDL Number: MFCD00048673
Molecular Formula: C26H52O2
Molecular Weight: 396.70
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 310
Melting point (°C): 56


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Lignoceric Acid Ethyl Ester, Ethyl Tetracosanoate, Tetracosanoic Acid Ethyl Ester

Formula: C₂₆H₅₂O₂
MW: 396.70 g/mol
Boiling Pt: 310 °C
Melting Pt: 56 °C
MDL Number: MFCD00048673
CAS Number: 24634-95-5
https://us.vwr.com/stibo/web/std.lang.all/30/90/28973090.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR