TCIAL0108

L0108-25ML
TCL0108-025MLEA 132.48 USD
TCL0108-025ML
TCIAL0108
CAS Number: 61931-81-5
MDL Number: MFCD00036495
Molecular Formula: C9H16O3
Molecular Weight: 172.22
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Flash Point (°C): 112
Specific Gravity (20/20): 0.99

Other Product Names / Synonyms: Lactic Acid cis-3-Hexen-1-yl Ester
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR