2-Chloroethyl laurate ≥95.0%

Supplier: TCI America

Synonyms: Lauric Acid 2-Chloroethyl Ester2-Chloroethyl dodecanoate

L0106-25G
TCL0106-025GEA 596 USD
TCL0106-025G
2-Chloroethyl laurate ≥95.0%
2-Chloroethyl laurate

CAS Number: 64919-15-9
MDL Number: MFCD00059475
Molecular Formula: C14H27ClO2
Molecular Weight: 262.82
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 187
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: Lauric Acid 2-Chloroethyl Ester

Formula: C₁₄H₂₇ClO₂
MW: 262.82 g/mol
Boiling Pt: 187 °C
Density: 0.96 (20 °C)
MDL Number: MFCD00059475
CAS Number: 64919-15-9
https://us.vwr.com/stibo/web/std.lang.all/30/87/28973087.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR