TCIAL0043

L0043-25G
TCL0043-025GEA 131.36 USD
TCL0043-025G
TCIAL0043
CAS Number: 591-64-0
MDL Number: MFCD00045959
Molecular Formula: C5H8O3
Molecular Weight: 270.29
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 123

Other Product Names / Synonyms: 3-Acetylpropionic Acid Calcium Salt, Levulinic Acid Calcium Salt, 4-Oxovaleric Acid Calcium Salt
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR