DL-Leucyl-DL-valine
Supplier: TCI America
Synonyms:
H-DL-Leu-DL-Val-OH
L0041-5G
L0041-1G
TCL0041-005GEA
240
USD
TCL0041-005G
TCL0041-001G
DL-Leucyl-DL-valine
DL-Leucyl-DL-valine
CAS Number: 35436-83-0
MDL Number: MFCD00068371
Molecular Formula: C11H22N2O3
Molecular Weight: 230.31
Form: Crystal
Color: White
Other Product Names / Synonyms: H-DL-Leu-DL-Val-OH
Formula:
C₁₁H₂₂N₂O₃ MW: 230.31 g/mol |
MDL Number:
MFCD00068371 CAS Number: 35436-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...