D-Leucylglycylglycine ≥98.0% (by HPLC, titration analysis)

Synonyms: H-D-Leu-Gly-Gly-OH

L0031-100MG
TCL0031-100MGEA 149.94 USD
TCL0031-100MG
D-Leucylglycylglycine ≥98.0% (by HPLC, titration analysis)
D-Leucylglycylglycine

CAS Number: 18625-22-4
MDL Number: MFCD00021740
Molecular Formula: C10H19N3O4
Molecular Weight: 245.28
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -58 deg (C=2, H2O)


Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-D-Leu-Gly-Gly-OH

Formula: C₁₀H₁₉N₃O₄
MW: 245.28 g/mol
MDL Number: MFCD00021740
CAS Number: 18625-22-4
https://us.vwr.com/stibo/web/std.lang.all/30/40/28973040.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR