Sodium laurate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Lauric acid sodium saltSodium dodecanoate

L0016-25G
TCL0016-025GEA 83.89 USD
TCL0016-025G
Sodium laurate ≥97.0% (by titrimetric analysis)
Sodium laurate

CAS Number: 629-25-4
MDL Number: MFCD00041754
Molecular Formula: C12H24O2
Molecular Weight: 222.30
Purity/Analysis Method: 97.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Anionic Surfactants, Bioscience, Lipids
Other Product Names / Synonyms: Lauric Acid Sodium Salt

Formula: C₁₂H₂₃NaO₂
MW: 222.304 g/mol
MDL Number: MFCD00041754
CAS Number: 629-25-4
https://us.vwr.com/stibo/web/std.lang.all/30/28/28973028.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR