Calcium lactobionate hydrate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Lactobionic Acid Calcium Salt

L0006-25G
TCL0006-025GEA 33.48 USD
TCL0006-025G
Calcium lactobionate hydrate ≥97.0% (by titrimetric analysis)
Calcium lactobionate hydrate

CAS Number: 5001-51-4
MDL Number: MFCD00135895
Molecular Formula: C12H22O12
Molecular Weight: 754.65
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Specific rotation [a]20/D: 24 deg (C=6, H2O)


Product Categories: Bioscience, Sugar Acids, Disaccharides
Other Product Names / Synonyms: Lactobionic Acid Calcium Salt

Formula: C₁₂H₂₂O₁₂
MW: 754.65 g/mol
MDL Number: MFCD00135895
CAS Number: 5001-51-4
https://us.vwr.com/stibo/web/std.lang.all/30/19/28973019.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR