5-Ketohexanenitrile ≥98.0%

Supplier: TCI America

Synonyms: 4-Acetylbutyronitrile5-Oxohexanenitrile

K0029-25ML
TCK0029-025MLEA 424 USD
TCK0029-025ML
5-Ketohexanenitrile ≥98.0%
5-Ketohexanenitrile

CAS Number: 10412-98-3
MDL Number: MFCD00010671
Molecular Formula: C6H9NO
Molecular Weight: 111.14
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Flash Point (°C): 108
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: 4-Acetylbutyronitrile, 5-Oxohexanenitrile

Formula: C₆H₉NO
MW: 111.14 g/mol
Boiling Pt: 88 °C
Density: 0.96 (20 °C)
Flash Pt: 108 °C
MDL Number: MFCD00010671
CAS Number: 10412-98-3
https://us.vwr.com/stibo/web/std.lang.all/29/81/28972981.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR