(S)-(+)-Ketopinic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (S)-(+)-Ketopinic Acid

K0028-5G K0028-1G
TCK0028-5GEA 289.28 USD
TCK0028-5G TCK0028-001G
(S)-(+)-Ketopinic acid ≥98.0% (by GC, titration analysis)
(S)-(+)-Ketopinic acid

CAS Number: 40724-67-2
MDL Number: MFCD00168053
Molecular Formula: C10H14O3
Molecular Weight: 182.22
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 235
Specific rotation [a]20/D: 25.5 deg (C=0.65, MeOH)

Formula: C₁₀H₁₄O₃
MW: 182.22 g/mol
Melting Pt: 235 °C
MDL Number: MFCD00168053
CAS Number: 40724-67-2
https://us.vwr.com/stibo/web/std.lang.all/29/80/28972980.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR