Calcium-3-methyl-2-oxobutyrate hydrate ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
3-Methyl-2-oxobutyric Acid Calcium Salt
K0021-5G
TCK0021-005GEA
167
USD
TCK0021-005G
Calcium-3-methyl-2-oxobutyrate hydrate ≥97.0% (by titrimetric analysis)
Calcium-3-methyl-2-oxobutyrate hydrate
MDL Number: MFCD00002580
Molecular Formula: C5H8O3
Molecular Weight: 270.29
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: 3-Methyl-2-oxobutyric Acid Calcium Salt
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...