1-[(2S)-3-(Acetylthio)-2-methylpropionyl]-L-proline hydrate ≥90.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1-[(2S)-3-(Acetylthio)-2-methyl-1-oxopropyl]-L-proline1-[(S)-3-(Acetylthio)isobutyryl]-L-proline

A1528-5G
TCA1528-005GEA 97.39 USD
TCA1528-005G
1-[(2S)-3-(Acetylthio)-2-methylpropionyl]-L-proline hydrate ≥90.0% (by titrimetric analysis)
1-[(2S)-3-(Acetylthio)-2-methylpropionyl]-L-proline hydrate

CAS Number: 64838-55-7
MDL Number: MFCD01860852
Molecular Formula: C11H17NO4S
Molecular Weight: 259.32
Purity/Analysis Method: 90.0% (T)
Form: Crystal
Specific rotation [a]20/D: -160 deg (C=1, MeOH)


Other Product Names / Synonyms: 1-[(S)-3-(Acetylthio)isobutyryl]-L-proline, 1-[(2S)-3-(Acetylthio)-2-methyl-1-oxopropyl]-L-proline

Formula: C₁₁H₁₇NO₄S
MW: 259.32 g/mol
MDL Number: MFCD01860852
CAS Number: 64838-55-7
https://us.vwr.com/stibo/web/std.lang.all/96/16/28929616.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR