5-(Isopropylamino)pentanol ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 5-(Isopropylamino)pentan-1-ol

I0757-1G
TCI0757-1GEA 82.18 USD
TCI0757-1G
5-(Isopropylamino)pentanol ≥98.0% (by titrimetric analysis)
5-(Isopropylamino)pentanol

CAS Number: 40447-21-0
Molecular Formula: C8H19NO
Molecular Weight: 145.25
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Boiling point (°C): 125
Melting point (°C): 44

Formula: C₈H₁₉NO
MW: 145.25 g/mol
Boiling Pt: 125 °C
Melting Pt: 44 °C
MDL Number: MFCD14708174
CAS Number: 40447-21-0
https://us.vwr.com/stibo/web/std.lang.all/26/38/28972638.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR