(S)-(4-Isopropyloxazolin-2-yl)ferrocene ≥98.0% (by GC, titration analysis)

Supplier: TCI America

I0749-1G
TCI0749-1GEA 134.95 USD
TCI0749-1G
(S)-(4-Isopropyloxazolin-2-yl)ferrocene ≥98.0% (by GC, titration analysis)
(S)-(4-Isopropyloxazolin-2-yl)ferrocene

CAS Number: 162157-03-1
Molecular Formula: C16H19FeNO
Molecular Weight: 297.18
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Specific rotation [a]20/D: -135 deg (C=1, EtOH)

Formula: C₁₆H₁₉FeNO
MW: 297.18 g/mol
CAS Number: 162157-03-1
https://us.vwr.com/stibo/web/std.lang.all/26/28/28972628.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR