3-Iodo-p-anisidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
3-Iodo-4-methoxyaniline
I0716-5G
I0716-25G
TCI0716-5GEA
113
USD
TCI0716-5G
TCI0716-25G
3-Iodo-p-anisidine ≥98.0% (by GC, titration analysis)
3-Iodo-p-anisidine
CAS Number: 74587-12-5
MDL Number: MFCD06656566
Molecular Formula: C7H8INO
Molecular Weight: 249.05
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 74
Other Product Names / Synonyms: 4-Amino-2-iodoanisole
Formula:
C₇H₈INO MW: 249.0490 g/mol Melting Pt: 74 °C |
MDL Number:
MFCD06656566 CAS Number: 74587-12-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...