3-Iodophenylsulfur pentafluoride ≥93.0%
Supplier: TCI America
I0694-1G
TCI0694-1GEA
294
USD
TCI0694-1G
3-Iodophenylsulfur pentafluoride ≥93.0%
3-Iodophenylsulphur pentafluoride
CAS Number: 286947-67-9
MDL Number: MFCD03788513
Molecular Formula: C6H4F5IS
Molecular Weight: 330.05
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 57
Specific Gravity (20/20): 2.06
Storage Temperature: 0-10°C
Formula:
C₆H₄F₅IS Boiling Pt: 57 °C Density: 2.06 (20 °C) |
MDL Number:
MFCD03788513 CAS Number: 286947-67-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...