4-Iodo-1H-pyrazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Iodopyrazole

I0692-5G I0692-25G
TCI0692-5GEA 41.62 USD
TCI0692-5G TCI0692-25G
4-Iodo-1H-pyrazole ≥98.0% (by GC, titration analysis)
4-Iodo-1H-pyrazole

CAS Number: 3469-69-0
MDL Number: MFCD00005244
Molecular Formula: C3H3IN2
Molecular Weight: 193.98
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 109

Formula: C₃H₃IN₂
MW: 193.97 g/mol
Melting Pt: 109 °C
MDL Number: MFCD00005244
CAS Number: 3469-69-0
https://us.vwr.com/stibo/web/std.lang.all/25/60/28972560.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR